missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ly-6G Rat, APC, Clone: 1A8, enQuireBio™
Rat Monoclonal Antibody
Brand: EnquireBio QAB56-APC-25ug
This item is not returnable.
View return policy
Specifications
Ly-6G | |
Monoclonal | |
0.5mg/mL | |
10 mM NaH2PO4, 150 mM NaCl, 0.09% NaN3, 0.1% gelatin, pH7.2 | |
Ly6g | |
Protein A/G Purified | |
RUO | |
546644 | |
IgG2a κ |
Flow Cytometry | |
1A8 | |
APC | |
XP_006521677 | |
Rat | |
25 μg | |
Primary | |
Store at 2-4°C. |