missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Lycopersicon Esculentum (Tomato) Lectin (LEL, TL), DyLight 649
Used as marker of blood vessels and microglial cells in rodents and in neuroscience research
Brand: Invitrogen™ L32472
Additional Details : Weight : 0.00100kg
Description
- Lycopersicon esculentum (Tomato) Lectin (LEL, TL), DyLight 649, is a bright deep-red/far-red LEL-fluorophore conjugate. The tomato lectin LEL is widely used as marker of blood vessels and microglial cells in rodents and in neuroscience research. It is a valuable tool in studies of rodent tumor angiogenesis and the tracing of neovascular development in xenograft models and brain research.
- LEL-DyLight 649 can be applied directly to tissue sections, or it can be introduced through intravascular perfusion for fluorescence detection.
- Very bright DyLight 649 conjugate detected with traditional Cy5/deep-red filter set, Ex/Em: 655/670 nm
- Applications: immunofluorescence, glycobiology
- Sugar specificity: [GlcNAc] 1,3-N-acetylglucosamine
- Inhibiting/eluting sugar: chitin hydrolysate
- Blood group specificity: non-specific
- Unconjugated lectin molecular weight: 71 KDa.
- Immunofluorescence working concentration: 5–20 μg/mL
- LEL is a very stable single-subunit glycoprotein containing ∼50 percent arabinose and galactose and may form multimeric aggregates in solution. It is provided at a 1 mg/mL concentration in 10 mM HEPES, 0.15 M NaCl, pH 7.5, 0.08% sodium azide, 0.1 mM Ca2. If a precipitate forms upon long-term storage, warm to 37°C.
Specifications
1 mg | |
DyLight 649 | |
DyLight Fluors | |
Solution | |
DyLight™ |
Tomato | |
When stored at or below 2–8°C in the dark, the product is stable for 6 months after reciept. |
|
Approved for shipment at Room Temperature or on Wet Ice | |
Lectins |
For Research Use Only. Not for use in diagnostic procedures.