missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Lymphoprep™
Density gradient medium for the isolation of mononuclear cells

£139.05 - £710.10
Specifications
Cell Type | Mononuclear Cells |
---|---|
For Use With (Application) | Cell Isolation |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15257179
![]() |
STEMCELL Technologies
07861 |
6 x 500 mL |
£710.10
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17171036
![]() |
STEMCELL Technologies
07801 |
250 mL |
£139.05
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17181036
![]() |
STEMCELL Technologies
07811 |
500 mL |
£383.40
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Lymphoprep™ is a density gradient medium recommended for the isolation of mononuclear cells from peripheral blood, cord blood, and bone marrow by exploiting differences in cell density
- Granulocytes and erythrocytes have a higher density than mononuclear cells and therefore sediment through the Lymphoprep™ layer during centrifugation
- Lymphoprep™ can be substituted for Ficoll-Paque™ without any need to change your existing protocols and is fully compatible with both SepMate™ and RosetteSep™
- Lymphoprep™ has a density of 1.077 g/mL
Specifications
Mononuclear Cells | |
Cell Isolation |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
STEMCELL Technologies Lymphoprep™