Learn More
Labconco™ Lyph-Lock™ Flask Top
Three-piece flasks for freeze drying samples. Provides a high-vacuum seal for efficient lyophilization.
£118.00 - £207.00
Product Code | Brand | Capacity (Metric) | Standard Taper Joint | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Standard Taper Joint | Price | Quantity & Availability | |||||
15143045
|
Labconco™
7556000 |
25 mL, 50 mL | 20/40 |
£129.00
Each |
Estimated Shipment: 11-06-2025
Log in to see stock. |
|||||
15153045
|
Labconco™
7556200 |
100 mL, 250 mL | 20/40 |
£189.00
Each |
Estimated Shipment: 11-06-2025
Log in to see stock. |
|||||
15163045
|
Labconco™
7556400 |
500 mL, 750 mL, 1000 mL | 20/40 |
£207.00
Each |
Estimated Shipment: 11-06-2025
Log in to see stock. |
|||||
15173035
|
Labconco™
7552000 |
25 mL, 50 mL | 19/38 |
£118.00
Each |
Estimated Shipment: 11-06-2025
Log in to see stock. |
|||||
15183035
|
Labconco™
7552200 |
100 mL, 250 mL | 19/38 |
£166.00
Each |
Estimated Shipment: 11-06-2025
Log in to see stock. |
|||||
15193035
|
Labconco™
7552400 |
500 mL, 750 mL, 1000 mL | 19/38 |
£184.00
Each |
Estimated Shipment: 11-06-2025
Log in to see stock. |
|||||
Description
Select your Lyph-Lock Flasks based on your sample sizes. Flasks should be filled to no more than one-third of their volume so that maximum surface area is achieved and efficient lyophilization is ensured.
- Flexible, silicone rubber seal grasps top and bottom and holds them securely together during pre-freezing and lyophilization
- A complete Lyph-Lock Flask includes a glass top and bottom and a rubber ring seal
- Top and bottom are high-strength borosilicate glass
- Seal is a flexibile, non-contaminating silicone rubber
- Tops, bottoms and seals are available separately as replacement components
- Adapters are required to attach flasks to freeze dry valve ports (sold separately)
- Borosilicate glass has maximum temperature is 490° C and silicone maximum temperature is 450° C
- Flat bottom reduces spills and provides better balance
- Compatible with all major brands of laboratory freeze dryers
- Entire flask may be autoclaved
Specifications
Borosilicate Glass |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Labconco™ Lyph-Lock™ Flask Top