Learn More
Macherey-Nagel™ Lyse Buffer RLY
A lyse buffer for use with NucleoSpin™ DNA RapidLyse and NucleoSpin™ 96 DNA RapidLyse to obtain purified genomic DNA. A 70 mL bottle is sufficient for 4 x 96 preps with the 96 DNA RapidLyse kit or 250 prep DNA RapidLyse kit.
Brand: Macherey-Nagel™ 740101.70
Additional Details : Weight : 0.07000kg
Description
The isolation of genomic DNA (gDNA) can be a time-consuming task. The NucleoSpin™ DNA RapidLyse kit is designed with a unique buffer chemistry to minimize the expenditure of time, work, and money.
- Unique lysis chemistry to efficiently release gDNA from cells, tissues, and organs
- Powerful lysis in one hour or less
- Superior gDNA yields compared to standard extraction methods
- Highest cost-efficiency of gDNA purification products
Specifications
Colorless | |
Dodecyl sulfate, sodium salt (2.5 - 3% by weight); Water | |
Sample lysis for genomic DNA isolation | |
Lyse Buffer | |
Liquid |
1.01 g/cm3 | |
NucleoSpin DNA RapidLyse or NucleoSpin 96 DNA RapidLyse kits | |
8.5 - 9.5 | |
70 mL |