missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Lysine (K)-specific Demethylase 4C/KDM4C/JMJD2C Recombinant Protein Antigen
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Brand: Novus Biologicals™ NBP2-57484PEP
Additional Details : Weight : 0.00970kg
Description
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human Lysine (K)-specific Demethylase 4C/KDM4C/JMJD2C. Source: E.coli Amino Acid Sequence: ARFSTASDMRFEDTFYGADIIQGERKRQRVLSSRFKNEYVADPVYRTFLKSSFQKKCQK The Lysine (K)-specific Demethylase 4C/KDM4C/JMJD2C Recombinant Protein Antigen is derived from E. coli. The Lysine (K)-specific Demethylase 4C/KDM4C/JMJD2C Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.Specifications
23081 | |
Lysine (K)-specific Demethylase 4C/KDM4C/JMJD2C Recombinant Protein Antigen | |
PBS and 1M Urea, pH 7.4. | |
EC 1.14.11, EC 1.14.11.-, GASC-1 protein, GASC1JmjC domain-containing histone demethylation protein 3C, Gene amplified in squamous cell carcinoma 1 protein, JHDM3C, JMJD2CbA146B14.1, jumonji domain containing 2C, Jumonji domain-containing protein 2C, KIAA | |
Unlabeled | |
100μL | |
E.coli |
>80% by SDS-PAGE and Coomassie blue staining | |
Store at −20°C. Avoid freeze-thaw cycles. | |
Blocking/Neutralizing, Control | |
KDM4C | |
Recombinant Protein Antigen | |
RUO | |
This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-53151. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml |
For Research Use Only.