Learn More
Novus Biologicals™ Lysine (K)-specific Demethylase 5B/KDM5B/JARID1B Antibody Blocking Peptide
Blocking Peptide
Brand: Novus Biologicals™ NBP1-97310PEP
Additional Details : Weight : 0.00970kg
Specifications
10765 | |
Store at -80°C. Avoid freeze-thaw cycles. | |
CT31FLJ10538, EC 1.14.11, EC 1.14.11.-, homolog 1A, JARID1BFLJ16281, jumonji, AT rich interactive domain 1B, Jumonji, AT rich interactive domain 1B (RBP2-like), lysine (K)-specific demethylase 5B, PLU-1FLJ12459, PLU1FLJ23670, PUT1, RBBP2H1, RBBP2H1AFLJ12491, RBP2-H1, Retinoblastoma-binding protein 2 homolog 1 | |
0.1 mg | |
Peptide dissolved in dH20. Contains no BSA. |
1 mg/mL | |
This peptide is useful as a blocking peptide for NBP1-97310. For further blocking peptide related protocol, click here . | |
KDM5B | |
Cancer, Epigenetics |