missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Lysine (K)-specific Demethylase 5B/KDM5B/JARID1B Recombinant Protein Antigen
Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition
Brand: Novus Biologicals™ NBP2-58615PEP
Additional Details : Weight : 0.00970kg
Description
A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human Lysine (K)-specific Demethylase 5B/KDM5B/JARID1B. Source: E.coli Amino Acid Sequence: PARRAKRMRAEAMNIKIEPEETTEARTHNLRRRMGCPTPKCENEKEMKSSIKQEPIERKDYIVENEKEKPKSRSKKATNAVDLYVCLLCGSGNDEDRL The Lysine (K)-specific Demethylase 5B/KDM5B/JARID1B Recombinant Protein Antigen is derived from E. coli. The Lysine (K)-specific Demethylase 5B/KDM5B/JARID1B Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.Specifications
10765 | |
Lysine (K)-specific Demethylase 5B/KDM5B/JARID1B Recombinant Protein Antigen | |
PBS and 1M Urea, pH 7.4. | |
CT31FLJ10538, EC 1.14.11, EC 1.14.11.-, homolog 1A, JARID1BFLJ16281, jumonji, AT rich interactive domain 1B, Jumonji, AT rich interactive domain 1B (RBP2-like), lysine (K)-specific demethylase 5B, PLU-1FLJ12459, PLU1FLJ23670, PUT1, RBBP2H1, RBBP2H1AFLJ124 | |
Unlabeled | |
100μL | |
E.coli |
>80% by SDS-PAGE and Coomassie blue staining | |
Store at −20°C. Avoid freeze-thaw cycles. | |
Blocking/Neutralizing, Control | |
KDM5B | |
Recombinant Protein Antigen | |
RUO | |
This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-51876. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml |
For Research Use Only.