missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Lysis/Binding Buffer for Dynabeads™ mRNA Purification Kits
Click to view available options
Quantity:
1 x 60 mL
Unit Size:
60mL
Description
This Lysis/Binding Buffer is included in the following Dynabeads™ mRNA Purification kits (Cat. No. 61011, 61012, and 61021). It is made available separately for applications that require more Lysis/Binding Buffer than is provided in the kit.
Specifications
Specifications
| Content And Storage | 1 x 60 mL, store (at 2°C to 8°C) |
| Product Line | Dynabeads |
| Quantity | 1 x 60 mL |
| Shipping Condition | Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction