Learn More
Macherey-Nagel™ Lysis Buffer FLB
Lysis buffer for breaking down the cell membrane in order to isolate the genomic DNA of a forensic sample (e.g., blood spot, buccal swab, cells, tissue, and many other sources) using the NucleoSpin™ 8/96 DNA Trace kit.
Brand: Macherey-Nagel™ 740322.500
Additional Details : Weight : 0.50000kg
Description
NucleoSpin™ 8 / 96 DNA Trace Kit is used for the isolation of genomic DNA from forensic samples utilizing either a flexible 8-well strip format for varying throughput or a proven 96-well plate format for high throughput.
- Time-saving parallel isolation of genomic DNA from forensic samples (e.g., buccal swabs, dried blood spots, cigarette filters)
- Processing possible under vacuum or by centrifugation
- Suitable for manual and automated processing
- Innovative MN Wash Plate minimizes risk of cross-contamination
- DNA ready to use for any kind of enzymatic reaction, e.g., STR analysis
- To be used for research purposes only
Specifications
Colorless | |
7.5 to 8.5 | |
50-01-1 | |
Room Temperature (18°C to 25°C) | |
Isolation of genomic DNA from forensic samples |
NucleoSpin 8/96 Trace Kits | |
500 mL | |
Odorless | |
Liquid | |
Guanidine hydrochloride |