Learn More
LysoPC(14:0/0:0), MedChemExpress
MedChemExpress LysoPC(14:0/0:0) is a lysophospholipid (LyP). It is a monoglycerophospholipid in which a phosphorylcholine moiety occupies a glycerol substitution site. LysoPC(14:0/0:0) has potent antispasmodic effect.
Brand: Medchem Express HY-113123
This item is not returnable.
View return policy
Description
LysoPC(14:0/0:0) is a lysophospholipid (LyP). It is a monoglycerophospholipid in which a phosphorylcholine moiety occupies a glycerol substitution site. LysoPC(14:0/0:0) has potent antispasmodic effect.Specifications
LysoPC(14:0/0:0) | |
White | |
C22H46NO7P | |
Ethanol : 50 mg/mL (106.93 mM; Need ultrasonic) | |
467.58 | |
98.0% | |
Solid |
20559-16-4 | |
Research | |
5 mg | |
CCCCCCCCCCCCCC(OC[C@@H](O)COP(OCC[N+](C)(C)C)([O-])=O)=O | |
467.58 | |
Research | |
Metabolism-sugar/lipid metabolism |
Research purposes only