missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LZIC Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
Antigen | LZIC |
---|---|
Concentration | 0.4 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16853234
|
Proteintech
14543-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16843234
|
Proteintech
14543-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LZIC belongs to the CTNNBIP1 family. It is ubiquitously expressed with highest levels in kidney. LZIC is up-regulated in several cases of gastric cancers.Specifications
LZIC | |
Western Blot | |
Unconjugated | |
Rabbit | |
Rat, Mouse, Human | |
Q5PQN7, Q8K3C3, Q8WZA0 | |
366507, 69151, 84328 | |
LZIC Fusion Protein Ag6047 | |
Primary | |
-20°C | |
LZIC |
0.4 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
LZIC, Protein LZIC | |
Lzic | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |