Learn More
IKA™ M 20.1 Grinding Chamber
Ensure effective processing with a second grinding chamber. IKA™ M 20.1 Grinding Chamber is designed for use with the IKA M 20 Universal Mill.
Brand:  IKA™ 0008006200
3920.68 GBP valid until 2025-03-31
Use promo code "24119" to get your promotional price.
Product Code. 10238661
  • £4466.00 / Each
Estimated Shipment: 26-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • The grinding chambers can be placed on the drive alternately
  • One chamber is cleaned and filled while the other is being processed
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Grinding Chamber
M20 Mill
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
IKA™ M 20.1 Grinding Chamber > For Use With: M20 Mill