Learn More
Gilson™ M-384 Well Adapter with Positioning by Hand
An accessory allowing for the use of 384-well microplates with positioning done by hand.
Brand: Gilson™
Additional Details : Weight : 0.90900kg
Description
This adapter is an accessory for the PLATEMASTER™ that allows use of 384-well microplates with positioning done by hand.
Specifications
Pipetter Accessory | |
ISO 9001 | |
Polypropylene | |
PLATEMASTER™ |
Not Autoclavable | |
Gray | |
Liquid Handling | |
1 Year |