Learn More
m-Anisoyl chloride, 99%, Thermo Scientific Chemicals
£25.30 - £338.00
Chemical Identifiers
CAS | 1711-05-3 |
---|---|
Molecular Formula | C8H7ClO2 |
Molecular Weight (g/mol) | 170.6 |
MDL Number | MFCD00000673 |
InChI Key | RUQIUASLAXJZIE-UHFFFAOYSA-N |
Synonym | m-anisoyl chloride, benzoyl chloride, 3-methoxy, m-methoxybenzoyl chloride, 3-anisoyl chloride, m-anisic acid chloride, 3-methoxy-benzoyl chloride, benzoyl chloride, m-methoxy, 3-methoxybenzoic acid chloride, m-anisoylchloride, 3-anisoylchlorid |
PubChem CID | 74374 |
IUPAC Name | 3-methoxybenzoyl chloride |
SMILES | COC1=CC=CC(=C1)C(=O)Cl |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10482211
|
Thermo Scientific Acros
212970050 |
5 g | Glass bottle |
£25.30
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10072770
|
Thermo Scientific Acros
212970250 |
25 g | Glass bottle |
£80.90
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10713154
|
Thermo Scientific Acros
212971000 |
100 g | Glass bottle |
£338.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1711-05-3 | |
170.6 | |
RUQIUASLAXJZIE-UHFFFAOYSA-N | |
74374 | |
COC1=CC=CC(=C1)C(=O)Cl |
C8H7ClO2 | |
MFCD00000673 | |
m-anisoyl chloride, benzoyl chloride, 3-methoxy, m-methoxybenzoyl chloride, 3-anisoyl chloride, m-anisic acid chloride, 3-methoxy-benzoyl chloride, benzoyl chloride, m-methoxy, 3-methoxybenzoic acid chloride, m-anisoylchloride, 3-anisoylchlorid | |
3-methoxybenzoyl chloride |
Specifications
1711-05-3 | |
1.2140g/mL | |
92°C | |
99% | |
CH3OC6H4COCl | |
MFCD00000673 | |
1.214 | |
Solubility in water: decomposes | |
COC1=CC=CC(=C1)C(=O)Cl | |
170.6 | |
170.6 | |
Liquid |
Colorless to Yellow | |
123°C to 125°C (15.0 mmHg) | |
Authentic | |
C8H7ClO2 | |
1.5575 to 1.5595 | |
10, 140 | |
m-anisoyl chloride, benzoyl chloride, 3-methoxy, m-methoxybenzoyl chloride, 3-anisoyl chloride, m-anisic acid chloride, 3-methoxy-benzoyl chloride, benzoyl chloride, m-methoxy, 3-methoxybenzoic acid chloride, m-anisoylchloride, 3-anisoylchlorid | |
RUQIUASLAXJZIE-UHFFFAOYSA-N | |
3-methoxybenzoyl chloride | |
74374 | |
99% | |
m-Anisoyl chloride |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 216-975-2
RUO – Research Use Only