missing translation for 'onlineSavingsMsg'
Learn More
m-Nitroacetophenone, 98%, Thermo Scientific Chemicals Product Code.: 10503681

m-Nitroacetophenone, 98%, Thermo Scientific Chemicals

Product Code. 10503681
500 g, Plastic bottle
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy

Product Code. 10503681

Brand: Thermo Scientific Acros 128335000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 121-89-1
Molecular Formula C8H7NO3
Molecular Weight (g/mol) 165.15
MDL Number MFCD00007259
InChI Key ARKIFHPFTHVKDT-UHFFFAOYSA-N
Synonym 3'-nitroacetophenone, 3-nitroacetophenone, m-nitroacetophenone, 1-3-nitrophenyl ethanone, ethanone, 1-3-nitrophenyl, acetophenone, 3'-nitro, m-acetylnitrobenzene, usaf ma-1, methyl 3-nitrophenyl ketone, 3-nitroacetofenon
PubChem CID 8494
IUPAC Name 1-(3-nitrophenyl)ethanone
SMILES CC(=O)C1=CC(=CC=C1)[N+](=O)[O-]

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 76.0°C to 79.0°C
Boiling Point 201.0°C to 203.0°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Plastic bottle
Linear Formula O2NC6H4COCH3
Quantity 500 g
Beilstein 07, 288
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol
Formula Weight 165.15
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material m-Nitroacetophenone, 98%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0467803 Certificate TypeCertificate of Analysis Date30/01/2025 Catalog Number128331000,128335000
Lot NumberA0395350 Certificate TypeCertificate of Analysis Date15/12/2022 Catalog Number128331000,128335000
Lot NumberA0446617 Certificate TypeCertificate of Analysis Date24/10/2022 Catalog Number128331000,128335000
Lot NumberA0434364 Certificate TypeCertificate of Analysis Date28/09/2021 Catalog Number128331000,128335000
Lot NumberA0425739 Certificate TypeCertificate of Analysis Date05/01/2021 Catalog Number128331000,128335000
Product Identifier
  • m-Nitroacetophenone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
m-Nitroacetophenone, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.