missing translation for 'onlineSavingsMsg'
Learn More

m-Phenetidine, 98%, Thermo Scientific Chemicals

Product Code. 10656544
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 621-33-0
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.18
MDL Number MFCD00007785
InChI Key WEZAHYDFZNTGKE-UHFFFAOYSA-N
Synonym m-phenetidine, benzenamine, 3-ethoxy, m-ethoxyaniline, 3-ethoxybenzenamine, unii-xe9kkl972r, ccris 4696, xe9kkl972r, 3-ethoxyphenylamine, 3-ethoxy aniline, 3-ethoxy-phenylamine
PubChem CID 12120
IUPAC Name 3-ethoxyaniline
SMILES CCOC1=CC=CC(N)=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Brown
Density 1.0300g/mL
Boiling Point 248.0°C
Flash Point >112°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula C2H5OC6H4NH2
Refractive Index 1.5650 to 1.5670
Quantity 5 mL
Specific Gravity 1.03
Formula Weight 137.18
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material m-Phenetidine, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'