missing translation for 'onlineSavingsMsg'
Learn More

m-Phenylenediamine, 99+%, Thermo Scientific Chemicals

Product Code. 10677672
Click to view available options
Quantity:
25 g
250 g
500 g
1 kg
5 kg
Packaging:
Glass bottle
Plastic drum
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-45-2
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.14
MDL Number MFCD00007799
InChI Key WZCQRUWWHSTZEM-UHFFFAOYSA-N
Synonym m-phenylenediamine, 1,3-phenylenediamine, 1,3-diaminobenzene, 1,3-benzenediamine, m-diaminobenzene, 3-aminoaniline, m-aminoaniline, meta-phenylenediamine, developer c, 3-phenylenediamine
PubChem CID 7935
ChEBI CHEBI:8092
IUPAC Name benzene-1,3-diamine
SMILES C1=CC(=CC(=C1)N)N

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 63.0°C to 65.0°C
Color Brown-Gray or Tan to Yellow
Boiling Point 282.0°C to 284.0°C
Flash Point 175°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula C6H4(NH2)2
Quantity 1 kg
Beilstein 03, IV, 79
Merck Index 15, 7395
Solubility Information Solubility in water: 350g/L (25°C). Other solubilities: soluble in alcohol,ether and chloroform
Formula Weight 108.14
Percent Purity 99+%
Physical Form Pellets
Chemical Name or Material m-Phenylenediamine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • m-Phenylenediamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'