Learn More
m-Toluidine, 99%, Thermo Scientific Chemicals
£21.20 - £468.23
Chemical Identifiers
CAS | 108-44-1 |
---|---|
Molecular Formula | C7H9N |
Molecular Weight (g/mol) | 107.16 |
MDL Number | MFCD00007808 |
InChI Key | JJYPMNFTHPTTDI-UHFFFAOYSA-N |
Synonym | m-toluidine, 3-toluidine, benzenamine, 3-methyl, 3-aminotoluene, m-methylaniline, m-tolylamine, m-toluidin, m-aminotoluene, m-methylbenzenamine, 3-methylbenzenamine |
PubChem CID | 7934 |
IUPAC Name | 3-methylaniline |
SMILES | CC1=CC=CC(N)=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10307670
|
Thermo Scientific Acros
157870050 |
5 mL | Glass Bottle |
£21.20
5mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10626172
|
Thermo Scientific Acros
157870010 |
1 L | Glass Bottle |
£102.00
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15109203
|
Thermo Scientific Acros
157870051 |
5 L | Plastic drum | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
108-44-1 | |
107.16 | |
JJYPMNFTHPTTDI-UHFFFAOYSA-N | |
7934 | |
CC1=CC=CC(N)=C1 |
C7H9N | |
MFCD00007808 | |
m-toluidine, 3-toluidine, benzenamine, 3-methyl, 3-aminotoluene, m-methylaniline, m-tolylamine, m-toluidin, m-aminotoluene, m-methylbenzenamine, 3-methylbenzenamine | |
3-methylaniline |
Specifications
108-44-1 | |
100.0 | |
White | |
203.0°C to 204.0°C | |
Authentic | |
C7H9N | |
1.5660 to 1.5680 | |
12,853 | |
0.98 | |
Solubility in water: 0.2 g/100mL (20°C). Other solubilities: >50 g/100mL methanol, >50 g/100mL ethanol, soluble in ether, dmf and diluted acids | |
CC1=CC=CC(N)=C1 | |
107.16 | |
107.15 | |
Crystalline Solid or Liquid |
98.5 | |
-30.0°C | |
0.9800g/mL | |
86°C | |
98.5% min. (GC) | |
CH3C6H4NH2 | |
MFCD00007808 | |
15,9696 | |
m-toluidine, 3-toluidine, benzenamine, 3-methyl, 3-aminotoluene, m-methylaniline, m-tolylamine, m-toluidin, m-aminotoluene, m-methylbenzenamine, 3-methylbenzenamine | |
JJYPMNFTHPTTDI-UHFFFAOYSA-N | |
3-methylaniline | |
7934 | |
99% | |
m-Toluidine, 99% |
Safety and Handling
GHS H Statement
May cause damage to organs through prolonged or repeated exposure.
Toxic if inhaled.
Toxic in contact with skin.
Very toxic to aquatic life.
Toxic if swallowed.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fre
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 203-583-1
missing translation for 'rtecsNumber' : XU2800000
missing translation for 'tsca' : TSCA
RUO – Research Use Only