missing translation for 'onlineSavingsMsg'
Learn More

m-Toluoyl chloride, 99%, Thermo Scientific Chemicals

Product Code. 10784081
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1711-06-4
Molecular Formula C8H7ClO
Molecular Weight (g/mol) 154.6
MDL Number MFCD00000681
InChI Key YHOYYHYBFSYOSQ-UHFFFAOYSA-N
Synonym m-toluoyl chloride, m-methylbenzoyl chloride, 3-toluoyl chloride, toluoyl chloride, benzoyl chloride, 3-methyl, m-toluyl chloride, meta methyl benzoyl chloride, 3-methylbenzoylchloride, 3-toluoylchloride, unii-w2vjw4350k
PubChem CID 74375
IUPAC Name 3-methylbenzoyl chloride
SMILES CC1=CC=CC(=C1)C(=O)Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Brown
Density 1.1700g/mL
Boiling Point 86.0°C (5.0 mmHg)
Flash Point 76°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula CH3C6H4COCl
Refractive Index 1.5475 to 1.5495
Quantity 5 g
Beilstein 09, 477
Specific Gravity 1.17
Solubility Information Solubility in water: decomposes.
Formula Weight 154.6
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material m-Toluoyl chloride, 99%
Show More Show Less
compliance-icons
Product Identifier
  • m-Toluoyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'