missing translation for 'onlineSavingsMsg'
Learn More

m-Tolylhydrazine hydrochloride, 98%, Thermo Scientific Chemicals

Product Code. 11400274
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 637-04-7
Molecular Formula C7H11ClN2
Molecular Weight (g/mol) 158.63
MDL Number MFCD00012932
InChI Key SHTBTGXNVWYODU-UHFFFAOYSA-N
Synonym 3-methylphenylhydrazine hydrochloride, m-tolylhydrazine hydrochloride, m-tolylhydrazinium 1+ chloride, 3-methylphenyl hydrazine hydrochloride, 3-methylphenylhydrazine hcl, m-tolyhydrazine hydrochloride, 1-3-methylphenyl hydrazine hydrochloride, 3-tolylhydrazine hydrochloride, 3-methyl phenylhydrazine hydrochloride, hydrazine, 3-methylphenyl-, monohydrochloride
PubChem CID 71562
IUPAC Name (3-methylphenyl)hydrazine;hydrochloride
SMILES [H+].[Cl-].CC1=CC=CC(NN)=C1

Specifications

Melting Point ∼193°C (decomposition)
Quantity 100 g
UN Number UN2811
Beilstein 3563995
Formula Weight 158.63
Percent Purity 98%
Chemical Name or Material m-Tolylhydrazine hydrochloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'