missing translation for 'onlineSavingsMsg'
Learn More

Grant Instruments™ Pump Plate for Heating Circulators

For use with Grant T100 Heating Circulator. Functional between –50 and 200°C

Brand:  Grant Instruments™ M-UC

Product Code. 15132605

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Metal pump inlet/outlet plate
  • 6.35mm
  • BSP/G1/4 female
  • Temp range -50 to 200°C
Specifications

Specifications

-50°C to +200°C
M-UC Pump Plate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers