missing translation for 'onlineSavingsMsg'
Learn More
Learn More
m-Xylene, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Description
m-Xylene is used in the production of isophthalic acid, which is a monomer and utilized in preparation of polyethylene terephthalate. It is used as an important starting material in the production of 2,4- and 2,6-xylidine. It is used as solvent in histology.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
m-Xylene is used in the production of isophthalic acid, which is a monomer and utilized in preparation of polyethylene terephthalate. It is used as an important starting material in the production of 2,4- and 2,6-xylidine. It is used as solvent in histology.
Solubility
Miscible with organic solvents. Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
m-Xylene is used in the production of isophthalic acid, which is a monomer and utilized in preparation of polyethylene terephthalate. It is used as an important starting material in the production of 2,4- and 2,6-xylidine. It is used as solvent in histology.
Solubility
Miscible with organic solvents. Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Melting Point | -47°C |
Density | 0.866 |
Boiling Point | 138°C to 139°C |
Quantity | 100 mL |
Flash Point | 25°C (77°F) |
Odor | Mild |
Refractive Index | 1.497 |
UN Number | UN1307 |
Beilstein | 605441 |
Merck Index | 14,10081 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'