Learn More
m-Xylene, 99%, Thermo Scientific Chemicals
£22.00 - £143.00
Chemical Identifiers
CAS | 108-38-3 |
---|---|
Molecular Formula | C8H10 |
Molecular Weight (g/mol) | 106.17 |
MDL Number | MFCD00008536 |
InChI Key | IVSZLXZYQVIEFR-UHFFFAOYSA-N |
Synonym | m-xylene, 1,3-dimethylbenzene, m-xylol, m-dimethylbenzene, meta-xylene, m-methyltoluene, 3-xylene, benzene, 1,3-dimethyl, 1,3-dimethylbenzol, santosol 150 |
PubChem CID | 7929 |
ChEBI | CHEBI:28488 |
IUPAC Name | 1,3-xylene |
SMILES | CC1=CC(C)=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11316656
|
Thermo Scientific Alfa Aesar
L03788.AE |
100 mL |
£22.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11326656
|
Thermo Scientific Alfa Aesar
L03788.AP |
500 mL |
£37.40
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11336656
|
Thermo Scientific Alfa Aesar
L03788.0F |
2500 mL |
£143.00
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
m-Xylene is used in the production of isophthalic acid, which is a monomer and utilized in preparation of polyethylene terephthalate. It is used as an important starting material in the production of 2,4- and 2,6-xylidine. It is used as solvent in histology.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applicationsm-Xylene is used in the production of isophthalic acid, which is a monomer and utilized in preparation of polyethylene terephthalate. It is used as an important starting material in the production of 2,4- and 2,6-xylidine. It is used as solvent in histology.
Solubility
Miscible with organic solvents. Immiscible with water.
Notes
Incompatible with strong oxidizing agents.
Chemical Identifiers
108-38-3 | |
106.17 | |
IVSZLXZYQVIEFR-UHFFFAOYSA-N | |
7929 | |
1,3-xylene |
C8H10 | |
MFCD00008536 | |
m-xylene, 1,3-dimethylbenzene, m-xylol, m-dimethylbenzene, meta-xylene, m-methyltoluene, 3-xylene, benzene, 1,3-dimethyl, 1,3-dimethylbenzol, santosol 150 | |
CHEBI:28488 | |
CC1=CC(C)=CC=C1 |
Specifications
-47°C | |
0.866 | |
25°C (77°F) | |
1.497 | |
MFCD00008536 | |
605441 | |
m-xylene, 1,3-dimethylbenzene, m-xylol, m-dimethylbenzene, meta-xylene, m-methyltoluene, 3-xylene, benzene, 1,3-dimethyl, 1,3-dimethylbenzol, santosol 150 | |
IVSZLXZYQVIEFR-UHFFFAOYSA-N | |
1,3-xylene | |
7929 | |
106.17 | |
m-Xylene |
108-38-3 | |
138°C to 139°C | |
Mild | |
C8H10 | |
UN1307 | |
14,10081 | |
Miscible with organic solvents. Immiscible with water. | |
CC1=CC(C)=CC=C1 | |
106.17 | |
CHEBI:28488 | |
99% |
Safety and Handling
GHS H Statement
H226-H312-H332-H315
Flammable liquid and vapor.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
P210-P233-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H226-H312+H332-H315-H319
missing translation for 'dotInformation' : Hazard Class: 3; Packaging Group: III
missing translation for 'einecsNumber' : 203-576-3
missing translation for 'rtecsNumber' : ZE2275000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only