missing translation for 'onlineSavingsMsg'
Learn More
Learn More
m-Xylylene dichloride, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
25 g
100 g
Description
m-Xylylene dichloride is used in pharmaceutical and used in medicine, brightener intermediates. α,α?-Dichloro-m-xylene is suitable for use in the synthesis of 1,7-bis(6-methoxy-2-formylphenyl)-1,7-dioxaheptane.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
m-Xylylene dichloride is used in pharmaceutical and used in medicine, brightener intermediates. α,α′-Dichloro-m-xylene is suitable for use in the synthesis of 1,7-bis(6-methoxy-2-formylphenyl)-1,7-dioxaheptane.
Solubility
Insoluble in water.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
m-Xylylene dichloride is used in pharmaceutical and used in medicine, brightener intermediates. α,α′-Dichloro-m-xylene is suitable for use in the synthesis of 1,7-bis(6-methoxy-2-formylphenyl)-1,7-dioxaheptane.
Solubility
Insoluble in water.
Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
Specifications
Specifications
Melting Point | 33°C to 37°C |
Density | 1.302 |
Boiling Point | 250°C to 255°C |
Flash Point | 144°C (291°F) |
Quantity | 25 g |
UN Number | UN2928 |
Beilstein | 2079898 |
Solubility Information | Insoluble in water. |
Formula Weight | 175.06 |
Percent Purity | 97% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'