missing translation for 'onlineSavingsMsg'
Learn More

m-Xylylene dichloride, 97%, Thermo Scientific Chemicals

Product Code. 11418790
Click to view available options
Quantity:
10 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

m-Xylylene dichloride is used in pharmaceutical and used in medicine, brightener intermediates. α,α?-Dichloro-m-xylene is suitable for use in the synthesis of 1,7-bis(6-methoxy-2-formylphenyl)-1,7-dioxaheptane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
m-Xylylene dichloride is used in pharmaceutical and used in medicine, brightener intermediates. α,α′-Dichloro-m-xylene is suitable for use in the synthesis of 1,7-bis(6-methoxy-2-formylphenyl)-1,7-dioxaheptane.

Solubility
Insoluble in water.

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-16-4
Molecular Formula C8H8Cl2
Molecular Weight (g/mol) 175.052
MDL Number MFCD00000912
InChI Key GRJWOKACBGZOKT-UHFFFAOYSA-N
Synonym 1,3-bis chloromethyl benzene, alpha,alpha'-dichloro-m-xylene, m-xylylene dichloride, benzene, 1,3-bis chloromethyl, m-bis chloromethyl benzene, m-xylylene chloride, m-dichloroxylene, benzene, 1,3-dichloromethyl, ccris 1774, m-xylene, .alpha.,.alpha.'-dichloro
PubChem CID 12275
IUPAC Name 1,3-bis(chloromethyl)benzene
SMILES C1=CC(=CC(=C1)CCl)CCl

Specifications

Melting Point 33°C to 37°C
Boiling Point 250°C to 255°C
Flash Point 144°C (291°F)
Quantity 10 g
UN Number UN2928
Beilstein 2079898
Formula Weight 175.06
Percent Purity 97%
Chemical Name or Material m-Xylylene dichloride

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'