missing translation for 'onlineSavingsMsg'
Learn More

Micronova™ M-Zone ScreenWipe™
BIOPHARMA

A low linting Sontara™ pre-saturated wipe designed for cleaning touch screens, impregnated with a blend of low ionic detergent and IPA.

Brand:  Micronova™ MZSW40-910


Product Code. 15521805

  • £222.00 / Pack of 200

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Cleans Monitors, Touch Screens, Lenses and Safety Glasses.
  • Combo pack. 2 individually packaged Sontara™ Wipes: 1 wet/ 1 dry
  • Wet wipe saturated with custom blend of Lab & Glass detergent and IPA (80/ 20%)
  • Removes Surface Contaminates
  • Cleans to a Streak Free Finish
  • Will Not Damage Coatings on Sensitive Screens and Lenses
  • Designed for Controlled Environments
  • A very low VOC content and is a gentler alternative to harsh ammonia based screen and instrument wipes available to the industry
  • Ideal in Defense/Aeronautics, Aerospace, Hi-tech Manufacturing and Clean Production
  • Specifications

    Specifications

    Pre-saturated Wipe
    22.9 cm
    20.3 cm
    White
    8 x 9 in.
    For lens and instrument screen cleaning.
    9 in.
    8 in.
    Polyester
    Wet and Dry screen wipes
    Individually Packaged
    Non-sterile
    Product Suggestions

    Product Suggestions

    Videos
    Safety and Handling

    Safety and Handling

    Product Identifier
    • M-Zone ScreenWipe

    Signal Word
    • Danger

    Hazard Category
    • LONG-TERM AQUATIC HAZARD Chronic 3
    • ACUTE AQUATIC Acute 1
    • Serious eye damage/eye irritation Category 2
    • Flammable solid Category 1
    • Specific target organ toxicity Category 3

    Hazard Statement
    • H228-Flammable solid.
    • H319-Causes serious eye irritation.
    • H336-May cause drowsiness or dizziness.
    • H412-Harmful to aquatic life with long lasting effects.

    Precautionary Statement
    • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
    • P280-Wear protective gloves/protective clothing/eye protection/face protection.
    • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

    Supplemental information
    • MIXTURE LIST-Contains : isoproyl alcohol

    SDS
    Documents

    Documents

    Certificates
    Special Offers

    Special Offers