Learn More
Thermo Scientific™ M110 Muffle Furnaces
Heat and dry metals at high temperatures with Thermo Scientific™ M110 Muffle Furnaces, that evenly distribute heat and operate economically in a small footprint.
Description
Description
Heat and dry metals at high temperatures with Thermo Scientific™ M110 Muffle Furnaces, that evenly distribute heat and operate economically in a small footprint.
Key Features:
  • 9L working volume
  • Two-shell design with air pocket between annealing box and outside walls
  • Multiple layers of high-quality, asbestos-free ceramic fiber insulation
  • Heating elements safely positioned in grooved blocks on side walls of the work space, covered with ceramic plates
  • Hinged door designed with parallel forced guidance system—hot surface always faces away from user when door is open
  • Adjustable upper limit cut-out protects samples and equipment
Ordering Information:
Models available with optional 24 hr. timer, Thermo Scientific™ Thermicon™ P digital program controller with microprocessor, exhaust fan and/or exhaust flue. Additional exhaust flue (50006394) and tray (50006408) available separately. Not available in North America.
Compatible with:
  • Exhaust flue (50006394)
  • Tray (50006408)