Learn More
Applied Biosystems™ M13 Human, Forward Primer, Desalted
M13 Human, Forward Primer, Desalted
Marke: Applied Biosystems™
Weitere Details : Weight : 0.00100kg
Beschreibung
Use our tool to find and order pre-designed primers for PCR and Sanger sequencing. Free of SNPs and dimers, target-specific, and suitable for universal PCR conditions, these primers can be ordered unmodified, M13-tailed, HPLC-purified, or desalted. All primers are checked by mass spectrometry.Spezifikation
M13 Human Forward Desalted Primer, 25 nmole, Solid, For PCR and Sanger Sequencing | |
Desalted | |
25 nmol | |
Solid |
M13 Human Forward DS Primer | |
Forward | |
HPLC |
For Research Use Only. Not for use in diagnostic procedures.