Learn More
Sartorius™ Infrared Moisture Analyzer
Designed for daily routine use in production or lab, quality control or inspection of incoming raw materials. Sartorius™ Infrared Moisture Analyzer has a rugged design and is user-friendly with a concise display that shows important information at a glance.
Brand:  Sartorius™ MA35M-000230V1
Code : 18
Additional Details : Weight : 7.60000kg
Product Code. 10601775
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
To ensure incredibly fast and effective sample heating the MA35 is equipped with two powerful metal rod dark infrared radiators. Easy to understand icons guide you in three steps from taring the sample pan to starting the measurement. All important operating parameters can be accessed and changed in seconds.
  • 40°C to 160°C temperature range, adjustable in increments of 1°C
  • Fully proven, reliable and effective dark infrared radiator for sample heating
  • Easy to operate by clear, easy to follow symbol guidance
  • Fully automatic end point detection or 0.1 to 99 minutes timer shut off mode
Specifications
Specifications
FDA/HACCP-Regulation
MA35M-000230V1
Metal Rod Heater
Digital
230 V