missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MabCaptureC Affinity Matrix

Thermo Scientific™ MabCaptureC Affinity Matrix is an agarose-based, high-binding-capacity Protein A chromatography resin with a rProtein A ligand produced in yeast.

Brand:  Thermo Scientific™ 1963662250

 View more versions of this product

Product Code. 30019889

This item is not returnable. View return policy

Description

Description

MabCaptureC Affinity Matrix is an agarose-based, high-binding-capacity Protein A chromatography resin with a rProtein A ligand produced in yeast. The engineered rProtein A binds at the CH2-CH3 interface of the Fc-region of IgGs and shows cross binding to the VH3 domain of IgGs.

MabCaptureC™ Affinity Matrix is suitable for the purification of monoclonal antibodies.The rProtein A ligand is immobilized on a highly cross-linked and uniform-sized agarose-based bead with an average particle size of 75 μm. The uniform particle size delivers superior performance characteristics compared to traditional Protein A resins.

Features of the MabCaptureC™ Affinity Matrix include:

  • High dynamic binding capacity for mAbs of >50 g/L at 4.8 min residence time
  • Purification of mAbs and a subpopulation of scFv's and Fab fragments containing VH3 binding domains
  • Clean and sanitize with standard sodium hydroxide agents for chromatography products
  • Excellent scalability
  • Non-animal derived

Main characteristics

  • Matrix: agarose-based, epoxide activated
  • Average particle size: 75 ± 10 μm
  • Ligand: recombinant C domain of Protein A
  • Ligand coupling method: epoxide coupling
  • Binding capacity: >50 g IgG/L resin at 4.8 min residence time and 10% breakthrough
  • Elution conditions: 0.1 M glycine pH 3.0, or 20–100 mM sodium citrate or acetate, pH 3.0
  • Cleaning conditions: 0.2 M sodium hydroxide for 15 minutes every cycle
  • Operating pressure: ≤3 bar
  • Pressure flow: >500 cm/h at 3 bar in a 2.6 x 20 cm column
  • Formulation buffer: 20% (v/v) ethanol

For process development and resin screening, MabCaptureC™ is also available in the following formats:

  • MabCaptureC™ MiniChrom Column, 1 mL (Cat. No. 5943662001)
  • MabCaptureC™ MiniChrom Column for process development, 5 mL (5943662005)
  • MabCaptureC™ RoboColumn, 200 μL (5943662200)
  • MabCaptureC™ RoboColumn, 600 μL (5943662600)
Specifications

Specifications

Affinity Matrix
Engineered rProtein A
75 ±10 μm
Suspension in 20% ethanol
250 mL
>50 g IgG/L
Agarose-based, epoxide-activated
CH2-CH3 interface Fc-region
Monoclonal antibody purification
Agarose Bead
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.