missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MabCaptureC™ RoboColumn™ Pre-packed Column

Product Code. p-9622009 Shop All Thermo Scientific Products
Click to view available options
Bed Volume:
200 μL
600 μL
Unit Size:
200µL
600µL
This item is not returnable. View return policy

Product Code. 30019462

Brand: Thermo Scientific™ 5943662200

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Thermo Scientific™ MabCaptureC™ RoboColumn™ are small chromatography columns used for fully automated and parallel chromatographic separations on robotic liquid handling workstations.

MabCaptureC™ RoboColumn™ are small chromatography columns used for fully automated and parallel chromatographic separations on robotic liquid handling workstations.

These pre-packed columns are ready to connect and have the following features:

  • Packed with MabCaptureC™ Affinity Matrix, offering high dynamic binding capacity and high resolution over a wide range of flow rates
  • Direct connection to standard chromatography systems

The 1 mL columns can be used for quick screenings of application feasibility and lab scale purification on a convenient and easy-to-use pre-packed column. The 10-cm bed height of the 5 mL column allows initial process development at a bench scale.

The pre-packed MabCaptureC™ Affinity Matrix contains engineered rProtein A that binds at the CH2-CH3 interface of the Fc-region of IgGs and shows cross binding to the VH3 domain of IgGs.

Features of MabCaptureC™ Affinity Matrix include:

  • High dynamic mAb binding capacity of > 50 g/L at 4.8 min residence time
  • Purification of mAbs and a subpopulation of scFv's and Fab fragments containing VH3 binding domains
  • Clean and sanitize with standard sodium hydroxide agents for chromatography products
  • Excellent scalability
  • Non-animal derived

Main resin characteristics:

  • Matrix: agarose-based, epoxide activated
  • Average particle size: 75 ± 10 μm
  • Ligand: recombinant C domain of Protein A
  • Ligand coupling method: epoxide coupling
  • Binding capacity: >55 g IgG/L resin at 5 min residence time and 10% breakthrough; >75 g IgG/L resin at 10 min. residence time and 10% breakthrough
  • Elution conditions: 0.1 M glycine pH 3.0, or 20–100 mM sodium citrate or acetate, pH 3.0
  • Cleaning conditions: 0.2 M sodium hydroxide for 15 minutes every cycle
  • Column dimensions: 0.8 x 2 cm (1 mL), 0.8 x 10 cm (5 mL)
  • Operating pressure: ≤ 3 bar
  • Formulation buffer: 20% (v/v) ethanol

Specifications

Bed Volume 200 μL
Binding Capacity >55 to 75 g IgG/L resin at 10% breakthrough (depending on residence time)
Diameter (Metric) 5 mm
Particle Size 75 ±10 μm
Length (Metric) 10 mm
Ligand Type Recombinant C domain of Protein A
Matrix Agarose-based, Epoxide-activated
Max. Operating Pressure ≤3 bar
Stationary Phase Agarose bead
Target IgG mAbs, and a subpopulation of scFv and Fab containing VH3 domain
Format 8-well strip
Quantity 200 μL
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.