Learn More
Thermo Scientific™ MAbPac™ HIC-10 HPLC Column
Separate monoclonal antibodies (mAbs) and related biologics with high resolution, excellent biocompatibility, and selectivity using the unique chemistry of this hydrophobic interaction column.
Brand: Thermo Scientific™ 088480
932.31 GBP valid until 2024-10-31
Use promo code "21692" to get your promotional price.
Description
Designed for separation of monoclonal antibodies (mAbs) and related biologics by hydrophobic interaction, the Thermo Scientific™ MAbPac™ HIC-10 LC column is a high resolution silica column. The MAbPac HIC-10 column's unique chemistry provides high resolution, excellent biocompatibility, and selectivity that is different from other HIC columns on the market.
Unique Chemistry Designed for Optimal Selectivity of mAbs
The MAbPac HIC-10 column is based on high-purity, spherical, wide-pore (1,000Å), 5μm silica particles functionalized with proprietary alkyl amide groups. Its advanced surface bonding technology leads to excellent chemical stability, high efficiency, and unique selectivity optimized for mAbs and different from other HIC phases.- Excellent biocompatibility and extremely low carryover
- High resolution and high efficiency
- Compatible with both organic solvent and aqueous mobile phases
- Rugged column stability
Specifications
Analytical Column | |
100 mm | |
5 μm | |
1000 Å | |
Silica | |
Spherical, Fully Porous, Ultrapure Silica | |
Hydrophobic Interaction |
4.6 mm | |
6000 psi (415 bar) | |
2 to 8 | |
60°C | |
MAbPac | |
Hydrophobic Interaction | |
Bio-LC |