Learn More
Thermo Scientific™ MAbPac™ Reversed Phase HPLC Columns
Use the new Thermo Scientific™ MAbPac™ RP columns for high-performance reversed-phase (RP) HPLC and LC/MS characterization of monoclonal antibodies (mAbs), fragments, variants, antibody drug conjugates (ADCs), PEGylated proteins, and bispecific proteins.
Brand: Thermo Scientific™ 303182
683.52 GBP valid until 2025-05-31
Use promo code "24127" to get your promotional price.
Description
Use the new Thermo Scientific™ MAbPac™ Reversed Phase columns for high-performance reversed-phase (RP) HPLC and LC/MS characterization of monoclonal antibodies (mAbs), fragments, variants, antibody drug conjugates (ADCs), PEGylated proteins, and bispecific proteins. These columns have excellent resolution, long column lifetime, and low carry-over. The surface chemistry, particle size, and pore size, have been optimized for high resolution separations.
Compatible with organic solvent used in HPLC and MS under RP conditions- Resin is based on a hydrophobic supermacroporous 4μm polymer particles
- Stable at extreme pH (0 to 14) and high temperature (up to 110 °C)
- 1,500Å large pore size polymeric resin
- Efficient separation of protein molecules with very low carry over
- Superior resolution power for monoclonal antibodies and related substances
- High efficiency with low carry-over
- Excellent MS compatibility
- Wide operating pH range: 0 to 14
- High temperature stability: up to 110°C
- High throughput

Specifications
Analytical Column | |
75 to 150 μL/min | |
4000 psi (275 bar) | |
1 to 7 | |
Each | |
PSDVB (Polystyrene Divinylbenzene) | |
PSDVB (Polystyrene Divinylbenzene) | |
Reversed Phase | |
Responsibly packaged |
1 mm | |
50 mm | |
4 μm | |
1500 Å | |
110°C | |
MAbPac | |
Reversed Phase | |
Bio-LC |
Product Suggestions
Customers who viewed this item also viewed.
Special Offers
-
Multiple Manufacturers
Chromatography and Chemicals Promotion
Explore our new promotion and get up to 70% off some of our most popular products.Promo Code: 24127Expires: 2025-05-31
New Offer Save Up to 70% -
Multiple Manufacturers
Elevate Your Beverage Testing with Precision Solutions
Get amazing savings on products for your analysesPromo Code: 24135Expires: 2025-07-13
New Offer Save Up to 85%
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.