Learn More
Thermo Scientific™ MacConkey Agar (EP, USP, JP, BP)
Detect, isolate and enumerate coliforms and intestinal pathogens in water, dairy products and biological specimens with this ready-to-use medium.
Brand: Thermo Scientific™ PO1142A
11.70 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Detect, isolate and enumerate coliforms and intestinal pathogens in water, dairy products and biological specimens with ready-to-use Thermo Scientific™ Oxoid™ MacConkey Agar (EP, USP, JP, BP). This differential medium gives a number of diagnostic indications such as bile tolerance, colony morphology and chromogenesis. MacConkey Agar corresponds to the medium recommended by the World Health Organizaton1, the Department of Health2 for the bacterial examination of water. The medium conforms to the formulations detailed in international pharmacopoeia3,4,5,6.
Use MacConkey Agar (EP, USP, JP, BP) for the cultivation of pathogens which may be present in a variety of specimens .
- Easy-to-read: easy to differentiate Escherichia coli (red non-mucoid colonies) from Staphyloccoccus spp. (pale pink opaque colonies) and Pseudomonas aeruginosa (green-brown) colonies.
- Recommended by World Health Organizaton1, the Department of Health 2
- Conforms to formulations detailed in International Pharmacopoeia3,4,5,6,
- Ready-to-use convenience of prepared media.
Due to its ability to support the growth of pathogenic Gram-positive cocci (e.g. staphylococci and enterococci) as well as Enterobacteriaceae, MacConkey Agar is particularly recommended for the cultivation of pathogens which may be present in a variety of specimens.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.

Specifications
Dark Red, clear | |
MacConkey Agar (EP, USP, JP, BP) | |
United Kingdom | |
19.5g ± 1.0g | |
10/Pk. |
2°C to 10°C | |
90mm Plate | |
Agar | |
Flow Wrapped - Single Layer |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
4500113 | Certificate of Analysis | 28/03/2025 | PO5002A,PO1142A |
4499217 | Certificate of Analysis | 24/03/2025 | PO5002A,PO1142A |
4496412 | Certificate of Analysis | 09/03/2025 | PO5002A,PO1142A |
4492467 | Certificate of Analysis | 23/02/2025 | PO5002A,PO1142A |
4489457 | Certificate of Analysis | 11/02/2025 | PO5002A,PO1142A |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.