missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MacConkey Agar No. 3

Detect and enumerate coliform organisms, such as E. coli, while isolating and detecting Salmonella and Shigella spp. in pathological and food specimens.

Brand:  Thermo Scientific™ PO0495A

11.49 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.


 View more versions of this product

Product Code. 12992618

  • £25.30 / Pack of 10
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Detect and enumerate coliform organisms, including E. coli, while isolating and detecting Salmonella and Shigella spp. in both pathological and food specimens using Thermo Scientific™ MacConkey Agar No. 3. One of the most famous formulations for detection of coliforms Salmonella and Shigella has improved its selectivity to Gram-positive organisms and is also a good medium for stool diagnostics.

Due to the inclusion of a specially prepared fraction of bile salts in addition to crystal violet, MacConkey Agar No. 3 is a highly selective medium for differentiating between coliforms and lactose and non-lactose fermenters while gram-positive cocci are completely inhibited.

Urinary tract infections, bacteremia, meningitis, and diarrheal disease are the most frequent clinical syndromes associated with Escherichia coli. Escherichia coli O157:H7 is associated with hemorrhagic colitis and hemolytic-uremic syndrome (HUS). Escherichia coli is an aerobic, gram-negative bacillus most frequently isolated from the intestines of humans and animals. As a result, it is used as an indicator of fecal contamination in water sources. This pathogen readily grows on a variety of culture media.

Salmonella
and Shigella are major causes of bacterial enteric illness. Transmission occurs via the fecal-oral route, person-to-person contact, or by the ingestion of contaminated food or water. A relatively small number of organisms ingested is capable of causing symptomatic infection. Salmonella and Shigella are aerobic, gram-negative, oxidase-negative bacilli. Some Salmonella and all Shigella are nonmotile. These organisms grow on standard media, such as MacConkey Agar, SS Agar, and XLD Agar. Enrichment broths should also be used when culturing for these organisms to allow for maximum recovery.

This product may not be available in all countries or territories. Please consult your local representative for availability.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Dark red, clear
90mm Monoplate
MacConkey Agar No. 3
19.5g ± 1.0g
10/Pk.
2°C to 10°C
United Kingdom
Agar
Flow Wrapped - Single Layer
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number4489754 Certificate TypeCertificate of Analysis Date14/02/2025 Catalog NumberPO0495A
Lot Number4488830 Certificate TypeCertificate of Analysis Date11/02/2025 Catalog NumberPO0495A
Lot Number4484198 Certificate TypeCertificate of Analysis Date20/12/2024 Catalog NumberPO0495A
Lot Number4482133 Certificate TypeCertificate of Analysis Date03/12/2024 Catalog NumberPO0495A
Lot Number4482141 Certificate TypeCertificate of Analysis Date03/12/2024 Catalog NumberPO0495A
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ MacConkey Agar No. 3 >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.