missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ MacConkey Agar without Salt
Click to view available options
Fill Volume:
19 mL
Fill Weight:
19g ± 5%
Unit Size:
Pack of 10
Description
Detect and enumerate coliform organisms in clinical samples with Thermo Scientific™ MacConkey Agar without Salt. The differential medium suppresses swarming of Proteus species and is, therefore, recommended for urine examination in the investigation of suspected urinary tract infection (UTI).
MacConkey Agar without Salt has the same formulation as MacConkey Agar except that it does not contain added salt and, therefore, provides a ‘low electrolyte medium’ on which most Proteus species do not spread. The medium has found particular favour for use in the examination of urine samples so that overgrowth of other organisms is prevented.
Specifications
Specifications
| Color | Salmon pink, transparent |
| Content And Storage | 2° to 10°C |
| Description | MacConkey Agar without Salt |
| Format | 90mm Monoplate |
| Manufacturing Origin | Germany |
| Product Type | Agar |
| Fill Volume | 19 mL |
| Fill Weight | 19g ± 5% |
| Packaging Type | Flow Wrapped - Single Layer |
| Quantity | 10/Pk. |
For In Vitro Diagnostic Use.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction