missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MacConkey Broth (Bottled)

Product Code. 12693438
This item is not returnable. View return policy

Product Code. 12693438

Brand: Thermo Scientific™ BO0550M

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Identify coliforms from water and milk samples with these ready-prepared bottles.

Identify coliforms from water and milk samples with differential Thermo Scientific™ Oxoid™ MacConkey Broth in ready-prepared bottles. MacConkey Broth is the standard medium for the primary isolation of coliform bacteria, and has been recommended for this purpose by the Public Health Laboratory Service Water Committee1 and the World Health Organization2.

Remove the need for autoclaving and dispensing with convenient, pre-filled and ready-to-use bottles of MacConkey Broth.

  • Convenient: pre-filled bottles are ready for immediate use.
  • Simple: easy-to-use with minimum training.
  • Long shelf life: up to 12 months.
  • Recommended by Public Health Laboratory Service Water Committee and World Health Organization.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific™ brand.

TRUSTED_SUSTAINABILITY

Specifications

Color Dark red, Clear
Content And Storage 2 to 25°C
Description MacConkey broth (Bottled)
Format Sirop Bottle
Manufacturing Origin United Kingdom
Product Type Agar
Fill Weight 100.0g - 101.5g
Packaging Type 10 sirop bottles with screw cap closures in a box
Quantity 10/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.