Learn More
Emma Safety Footwear Mace M Safety Shoes
A superb shoe for galvanizing and welding metal. Closed front flap protects against stray sparks and hot metal splinters. Hydro-Tec™ Sanitized Silver keep feet dry and fresh.
Brand: Emma Safety Footwear MM50486341
This item is not returnable.
View return policy
Description
- S3-Safety shoe with sole plate for wet work
- GripForce™ Duo / HRO
- Full grain leather
- Heat resistant outsole
- Hydro-Tec™ Sanitized Silver
Specifications
EN ISO 20345:2011 | |
Black | |
Hydro-Tec™ Sanitized Silver | |
41 |
Laces | |
Mace M | |
Leather | |
Steel |