missing translation for 'onlineSavingsMsg'
Learn More

Hellma™ Macro Cuvet Absorption Cell

Manufactured from Quartz Silica. Hellma™ Macro Cuvet Absorption Cell provides photometry with two windows, a 10mm path length and comes complete with lid.

Brand:  Hellma™ 100-10-40

Product Code. 10518691

  • £215.00 / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

For use with photometric analysis.
  • 10mm light path
  • 2 windows
  • Material code: QS, Quartz Silica
  • 12.5 x 12.5 x 45mm (W x D x H)
Specifications

Specifications

3500 μL
10 mm
12.5 mm
Quartz Silica
45 mm
Photometric Analysis
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.