missing translation for 'onlineSavingsMsg'
Learn More

Hellma Macro Cuvette, 111-OS

Macro-type cell made of special optical glass and used for fluorescence assays.

Brand:  Hellma 111-10-20

Product Code. 10363551

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

  • Optical glass construction
  • PTFE stopper included
  • VIS range
  • For fluorescence measurements
Specifications

Specifications

3500 μL
PTFE
Base Thickness: 1.25 mm
1/Pk
320 to 2500 nm
12.5 mm
Special Optical Glass
10 mm
PTFE Stopper
10 x 10 mm
46 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers