missing translation for 'onlineSavingsMsg'
Learn More

Cytiva (formerly Pall Lab) Macrosep™ Advance Centrifugal Devices with Omega Membrane

Macrosep Advance Centrifugal Devices with Omega Membrane quickly concentrates up to 20 mL of biological sample.

Brand:  Cytiva (formerly Pall Lab) MAP003C37

351.71 GBP valid until 2024-12-31
Use promo code "21930" to get your promotional price.



Additional Details : Weight : 1.11150kg

 View more versions of this product

Product Code. 17144731

  • £425.00 / Pack of 24
Estimated Shipment: 19-07-2024
to see stock.
Add to basket

Description

Description

  • Exchange buffer or remove salt of chromatography eluates and gradient fractions
  • For research applications only
Specifications

Specifications

Gray
PES
Non-sterile
5 to 20 mL (Sample)
EFA: 7.2 cm2
3 kDa
24/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers