missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva (formerly Pall Lab) Macrosep™ Advance Centrifugal Devices with Omega Membrane
Macrosep Advance Centrifugal Devices with Omega Membrane quickly concentrates up to 20 mL of biological sample.
Brand: Cytiva (formerly Pall Lab) MAP010C37
269.51 GBP valid until 2024-12-31
Use promo code "21930" to get your promotional price.
Description
- Exchange buffer or remove salt of chromatography eluates and gradient fractions
- For research applications only
Specifications
Blue | |
PES | |
Non-sterile | |
5 to 20 mL (Sample) |
EFA: 7.2 cm2 | |
10 kDa | |
24/Pk. |