missing translation for 'onlineSavingsMsg'
Learn More

Cytiva (formerly Pall Lab) Macrosep™ Advance Centrifugal Devices with Supor Membrane

Particulate removal for longer HPLC and UHPLC column life.

Brand:  Cytiva (formerly Pall Lab) MAPM02C68

Additional Details : Weight : 2.53650kg

 View more versions of this product

Product Code. 17134741

  • £1105.00 / Pack of 100
Estimated Shipment: 30-08-2024
to see stock.
Add to basket

Description

Description

  • High spin speed and larger EFA reduces spin times
  • For samples from 3 to 20 mL
  • For research applications only
Specifications

Specifications

Aqua
PES
Non-sterile
5 to 20 mL (Sample)
EFA: 7.2 cm2
0.2 μm
100/Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers