Learn More
MAD2L2 Mouse anti-Human, Unconjugated, Clone: 2A4C2, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67100-1-IG-150UL
 View more versions of this product
Product Code. 16819535
  • £349.00 / 150µL
Explore more special offers
Description
Description
The MAD2L2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human MAD2L2. This antibody recognizes human, mouse, rat antigen. The MAD2L2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
Specifications
MAD2L2
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunohistochemistry, ELISA
2A4C2
Unconjugated
150 μL
10459
IgG2b