Learn More
MAD2L2 Mouse anti-Human, Unconjugated, Clone: 2A4C2, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67100-1-IG-20UL
This item is not returnable.
View return policy
Description
The MAD2L2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human MAD2L2. This antibody recognizes human, mouse, rat antigen. The MAD2L2 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
MAD2L2 | |
Monoclonal | |
1000 μg/mL | |
Mouse | |
Primary | |
Human, Rat, Mouse |
Western Blot, Immunohistochemistry, ELISA | |
2A4C2 | |
Unconjugated | |
20 μL | |
10459 | |
IgG2b |