Learn More
MP Biomedicals™ Magbeads FastDNA Kit for FFPE
MagBeads FastDNA Kit for FFPE is intended for rapid extraction of DNA from FFPE sample. This kit uses two combination methods. High-salt Bind is conducive to remove pigments or polysaccharides from complex FFPE samples, so as to improve the purity of nucleic acid and avoid blocking aligent 2100.
Brand: MP Biomedicals™ 116576096
Additional Details : Weight : 0.52500kg
Description
MagBeads FastDNA Kit for FFPE is based on the purification method of high binding magnetic particles:
- The sample is lysed and digested
- DNA is released into the lysate
- After addition of magnetic particles and binding solution, DNA will be adsorbed on the surface of magnetic particles, and impurities such as proteins will be removed without adsorption
- The adsorbed particles were washed with washing buffer to remove the proteins and impurities, washed with ethanol to remove salts
- Finally the DNA was eluted with Elution Buffer
Specifications
Kit | |
Mini | |
DNA and RNA Sequencing |
Magnetic Bead | |
FFPE | |
96 Preps |