Learn More
MP Biomedicals™ MagBeads FastDNA Kit for Soil
MagBeads FastDNA Kit for Soil - Quick Isolation of genomic DNA from soil using magnetic beads technology.
Brand: MP Biomedicals™ 116561050
367.56 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : Weight : 0.80000kg
Description
- The MagBeads FastDNA™ Kit for Soil allows quick and efficient isolation of high-quality genomic DNA from soil in less than 60 mins
- Samples are placed into Lysing Matrix E tubes and used with FastPrep™ Instruments from MP Biomedicals to effectively lyse within 40 seconds:
- Host cells
- Bacteria
- Fungi
- Viruses
- Protists
- Other cells present in soil samples
- The kit is also compatible with most of the automated nucleic acid extraction instruments on the market or it can be operated manually. It consists of specially formulated reagents to eliminate:
- Humic acid
- Polysaccharides
- Phenolic compounds
- Enzyme inhibitors from soil
- It allows for extraction of highly pure genomic DNA ready for:
- PCR
- Restriction digestion
- Electrophoresis
- Other desired applications
Specifications
Kit | |
Soil (Various Types) | |
50 Preps |
Mini | |
DNA and RNA Sequencing |
Safety and Handling
- MagBeads FastDNA Kit for Soil
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H332-Harmful if inhaled.
Precautionary Statement
- P101-If medical advice is needed, have product container or label at hand.
- P102-Keep out of reach of children.
- P103-Read label before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P330-Rinse mouth.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Sodium Dodecyl Sulfate, Lithium Chloride, Disodium Dihydrogen Ethylenediaminetetraacetate, Aluminium