Learn More
MP Biomedicals™ Magbeads FastDNA/RNA Kit for Virus
MagBeads FastDNA/RNA Kit for Virus is intended for extracting total viral nucleic acid from cell-free/low-cell content biological samples such as body fluids, serum, plasma, soaking solution, tissue homogenate, and culture supernatant.
Brand: MP Biomedicals™ 116571192
Additional Details : Weight : 0.44500kg
Description
MagBeads FastDNA/RNA Kit for Virus is based on the purification method of high binding magnetic particles:
- The sample is lysed and digested with Protease
- DNA/RNA is released into the lysate
- After addition of magnetic particles and binding solution, DNA/RNA will be adsorbed on the surface of magnetic particles, and impurities such as proteins will be removed without adsorption
- The adsorbed particles are washed with washing solution to remove proteins and impurities, washed with ethanol to remove salts, and finally DNA/RNA is eluted with Nuclease Free Water
Specifications
Kit | |
Mini | |
DNA and RNA Sequencing |
Magnetic Bead | |
Virus | |
192 Preps |