Learn More
MP Biomedicals™ Magbeads FastRNA Kit for FFPE
MagBeads FastRNA Kit for FFPE is intended for rapid extraction of RNA from tissue, cells, blood, and other clinical samples. RNA can be used directly for RT-PCR, quantitative RT-PCR and so on.
Brand: MP Biomedicals™ 116573192
Additional Details : Weight : 1.18000kg
Description
MagBeads FastRNA Kit for FFPE is based on the purification method of high binding magnetic particles:
- The sample is lysed and digested
- RNA is released into the lysate
- After addition of magnetic particles and binding solution, RNA will be adsorbed on the surface of magnetic particles, and impurities such as proteins will be removed without adsorption
- The adsorbed particles were washed with washing buffer to remove the proteins and impurities, washed with ethanol to remove salts
- Finally the RNA was eluted with Elution Buffer
Specifications
Kit | |
Mini | |
DNA and RNA Sequencing |
Magnetic Bead | |
FFPE | |
192 Preps |